Assay Details
| Gene Symbol: Kif3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290805.1 | NP_001277734.1 | 5 - 6 | 778 | 130 | |
| NM_001290806.1 | NP_001277735.1 | 5 - 6 | 778 | 130 | ||
| NM_008443.4 | NP_032469.2 | 5 - 6 | 778 | 130 | ||
| GenBank mRNA | AK147078.1 | - | 5 - 6 | 728 | 130 | |
| AK161223.1 | - | 5 - 6 | 769 | 130 | ||
| AK161920.1 | - | 5 - 6 | 752 | 130 | ||
| AK169425.1 | - | 5 - 6 | 752 | 130 | ||
| BC023936.1 | - | 5 - 6 | 728 | 130 | ||
| BC044720.1 | - | 5 - 6 | 739 | 130 | ||
| BC052707.1 | - | 5 - 6 | 744 | 5702415 | 130 | |
| D12645.1 | - | 5 - 6 | 754 | 130 | ||
More Information
Set Membership: |
|

Kif3a
Genomic Map