Assay Details
| Gene Symbol: Il4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027491.1 | 1 - 2 | 262 | 116 | ||
| GenBank mRNA | AA199406.1 | - | 1 - 2 | 262 | 116 | |
| AF359558.1 | - | 1 - 2 | 201 | 116 | ||
| BX529138.1 | - | 1 - 2 | 271 | 116 | ||
More Information
Set Membership: |
|

Il4
Genomic Map