Assay Details
| Gene Symbol: Gm12216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033332.1 | 2 - 3 | 248 | 75 | ||
| GenBank mRNA | AK138811.1 | - | 2 - 3 | 249 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | interferon regulatory factor 1 | Irf1 |
AU020929, Irf-1 |
-35.2kb | Anti-sense | Chr. 11 - 53770014 - 53778374 on NCBI Build GRCm38 |
| Downstream |
solute carrier family 22 (organic cation transporter), member 5 |
Slc22a5 |
Lstpl, Octn2, jvs |
14.4kb | Sense | Chr. 11 - 53864542 - 53891799 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm12216
Genomic Map