Assay Details
| Gene Symbol: P4ha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136076.2 | NP_001129548.1 | 2 - 3 | 248 | 63 | |
| NM_011031.2 | NP_035161.2 | 2 - 3 | 248 | 63 | ||
| GenBank mRNA | AK053504.1 | - | 2 - 3 | 209 | 63 | |
| AK142624.1 | - | 2 - 3 | 197 | 63 | ||
| AK144124.1 | - | 2 - 3 | 194 | 63 | ||
| AK145535.1 | - | 2 - 3 | 175 | 63 | ||
| BC018411.1 | - | 2 - 3 | 160 | 3157481 | 63 | |
| U16163.1 | - | 1 - 2 | 135 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK145535.1, BC018411.1, NM_001136076.2, NM_011031.2, U16163.1 transcripts.
Set Membership: |
|

P4ha2
Genomic Map