| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100004977.mm.1 |
NM_001033597.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004977.mm.1 |
NM_001033597.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004977.mm.1 |
NM_001033598.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004977.mm.1 |
NM_001033598.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004977.mm.1 |
NM_001033599.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004977.mm.1 |
NM_001033599.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004977.mm.1 |
NM_144823.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004977.mm.1 |
NM_144823.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004977.mm.1 |
TC1100000621.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100004978.mm.1 |
NM_001033597.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004978.mm.1 |
NM_001033597.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004978.mm.1 |
NM_001033598.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004978.mm.1 |
NM_001033598.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004978.mm.1 |
NM_001033599.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004978.mm.1 |
NM_001033599.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004978.mm.1 |
NM_144823.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004978.mm.1 |
NM_144823.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004978.mm.1 |
TC1100000621.mm.1 |
MTA TC |