Assay Details
| Gene Symbol: Hnrnpab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048061.1 | NP_001041526.1 | 2 - 3 | 459 | 56 | |
| NM_010448.3 | NP_034578.1 | 2 - 3 | 459 | 56 | ||
| GenBank mRNA | AK013384.1 | - | 2 - 3 | 426 | 56 | |
| AK013709.1 | - | 425 | 56 | |||
| AK076132.1 | - | 2 - 3 | 422 | 56 | ||
| AK165609.1 | - | 3 - 4 | 406 | 56 | ||
| BC043069.1 | - | 2 - 3 | 413 | 6402930 | 56 | |
| CA325334.1 | - | 1 - 2 | 157 | 56 | ||
| D90151.1 | - | 2 - 3 | 414 | 56 | ||
| DQ397821.1 | - | 1 - 2 | 221 | 56 | ||
| DQ397822.1 | - | 1 - 2 | 221 | 56 | ||
| L36663.1 | - | 418 | 56 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Hnrnpab
Genomic Map