Assay Details
| Gene Symbol: Mapk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163672.1 | NP_001157144.1 | 7 - 8 | 953 | 86 | |
| NM_207692.2 | NP_997575.2 | 7 - 8 | 953 | 86 | ||
| GenBank mRNA | AF052466.1 | - | 6 - 7 | 689 | 86 | |
| AF052467.1 | - | 6 - 7 | 689 | 86 | ||
| AK031959.1 | - | 7 - 8 | 952 | 86 | ||
| AK145083.1 | - | 7 - 8 | 928 | 86 | ||
| BC028341.1 | - | 7 - 8 | 935 | 4192796 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mapk9
Genomic Map