Assay Details
| Gene Symbol: Cnot6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290741.1 | NP_001277670.1 | 7 - 8 | 876 | 90 | |
| NM_212484.2 | NP_997649.1 | 8 - 9 | 1230 | 90 | ||
| GenBank mRNA | AK129307.1 | - | 2 - 3 | 2168 | 90 | |
| AY043269.1 | - | 7 - 8 | 874 | 90 | ||
| BC023766.1 | - | 3 - 4 | 341 | 90 | ||
| BC049984.1 | - | 3 - 4 | 400 | 90 | ||
| BC057190.1 | - | 2 - 3 | 327 | 90 | ||
| BC062950.1 | - | 8 - 9 | 1218 | 6840162 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnot6
Genomic Map