Assay Details
| Gene Symbol: Cnot6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290741.1 | NP_001277670.1 | 8 - 9 | 1030 | 95 | |
| NM_212484.2 | NP_997649.1 | 9 - 10 | 1384 | 95 | ||
| GenBank mRNA | AK129307.1 | - | 3 - 4 | 2322 | 95 | |
| AY043269.1 | - | 8 - 9 | 1028 | 95 | ||
| BC023766.1 | - | 4 - 5 | 495 | 95 | ||
| BC049984.1 | - | 4 - 5 | 554 | 95 | ||
| BC057190.1 | - | 3 - 4 | 481 | 95 | ||
| BC062950.1 | - | 9 - 10 | 1372 | 6840162 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnot6
Genomic Map