Assay Details
| Gene Symbol: Clk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF033566.1 | - | 3 - 4 | 460 | 87 | |
| BC012675.1 | - | 3 - 4 | 482 | 3982920 | 87 | |
| CT010293.1 | - | 2 - 3 | 382 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clk4
Genomic Map