| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100024171.mm.1 |
NM_001024230.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024171.mm.1 |
NM_001024230.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024171.mm.1 |
TC1100002642.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100024170.mm.1 |
NM_001024230.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024170.mm.1 |
NM_001024230.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024170.mm.1 |
TC1100002642.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100024169.mm.1 |
NM_001024230.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024169.mm.1 |
NM_001024230.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024169.mm.1 |
TC1100002642.mm.1 |
MTA TC |