Assay Details
| Gene Symbol: Gm5431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024230.2 | NP_001019401.2 | 3 - 4 | 272 | 69 | |
| GenBank mRNA | AK144287.1 | - | 3 - 4 | 273 | 69 | |
| AK165745.1 | - | 3 - 4 | 273 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK144287.1, AK165745.1, NM_001024230.2 transcripts.
Set Membership: |
|

Gm5431
Genomic Map