Assay Details
| Gene Symbol: Med7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104556.1 | NP_001098026.1 | 1 - 2 | 232 | 141 | |
| GenBank mRNA | AK144918.1 | - | 1 - 2 | 190 | 141 | |
| BE333513.1 | - | 1 - 2 | 162 | 141 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK144918.1, NM_001104556.1 transcripts.
Set Membership: |
|

Med7
Genomic Map