Assay Details
| Gene Symbol: Med7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104530.1 | NP_001098000.1 | 1 - 2 | 59 | 69 | |
| GenBank mRNA | AK010357.1 | - | 1 - 2 | 55 | 69 | |
| BY135693.1 | - | 1 - 2 | 57 | 69 | ||
| CK030901.1 | - | 1 - 2 | 59 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK010357.1, NM_001104530.1 transcripts.
Set Membership: |
|

Med7
Genomic Map