Assay Details
| Gene Symbol: Ebf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290709.1 | NP_001277638.1 | 2 - 3 | 375 | 94 | |
| NM_001290710.1 | NP_001277639.1 | 2 - 3 | 375 | 94 | ||
| NM_001290711.1 | NP_001277640.1 | 2 - 3 | 375 | 94 | ||
| NM_007897.3 | NP_031923.1 | 2 - 3 | 375 | 94 | ||
| GenBank mRNA | AK031239.1 | - | 2 - 3 | 370 | 94 | |
| AK035781.1 | - | 2 - 3 | 258 | 94 | ||
| AK051438.1 | - | 2 - 3 | 374 | 94 | ||
| AK137389.1 | - | 2 - 3 | 297 | 94 | ||
| AK139021.1 | - | 2 - 3 | 373 | 94 | ||
| AK141175.1 | - | 2 - 3 | 375 | 94 | ||
| BC139362.1 | - | 2 - 3 | 336 | 8862384 | 94 | |
| BC139364.1 | - | 2 - 3 | 336 | 8862386 | 94 | |
| L12147.1 | - | 2 - 3 | 341 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001290711.1 transcripts.
Set Membership: |
|

Ebf1
Genomic Map