Assay Details
| Gene Symbol: Thg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080969.3 | NP_001074438.1 | 3 - 4 | 572 | 89 | |
| NM_001290737.1 | NP_001277666.1 | 4 - 5 | 515 | 89 | ||
| GenBank mRNA | AK010876.1 | - | 3 - 4 | 564 | 89 | |
| AK015467.1 | - | 3 - 4 | 366 | 89 | ||
| AK016076.1 | - | 4 - 5 | 516 | 89 | ||
| AK017532.1 | - | 3 - 4 | 555 | 89 | ||
| BC115560.1 | - | 3 - 4 | 560 | 40101901 | 89 | |
| BC115561.1 | - | 3 - 4 | 560 | 40101902 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Thg1l
Genomic Map