Assay Details
| Gene Symbol: Timd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178759.4 | NP_848874.3 | 2 - 3 | 412 | 82 | |
| GenBank mRNA | AK046560.1 | - | 2 - 3 | 411 | 82 | |
| AK079787.1 | - | 2 - 3 | 411 | 82 | ||
| AK080042.1 | - | 2 - 3 | 411 | 82 | ||
| AK080212.1 | - | 2 - 3 | 413 | 82 | ||
| AY376716.1 | - | 2 - 3 | 411 | 82 | ||
| AY376717.1 | - | 2 - 3 | 411 | 82 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BC032879.1 | - | 921 | Chr. - - [-] on Build | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Timd4
Genomic Map