Assay Details
| Gene Symbol: Pttg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001131054.1 | NP_001124526.1 | 5 - 6 | 580 | 62 | |
| GenBank mRNA | AF069051.1 | - | 4 - 5 | 815 | 62 | |
| AK002473.1 | - | 4 - 5 | 546 | 62 | ||
| AK008704.1 | - | 4 - 5 | 547 | 62 | ||
| AK088387.1 | - | 4 - 5 | 546 | 62 | ||
| BC023324.1 | - | 5 - 6 | 586 | 4946234 | 62 | |
| DV657646.1 | - | 4 - 5 | 550 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pttg1
Genomic Map