Assay Details
| Gene Symbol: Pttg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001131054.1 | NP_001124526.1 | 1 - 2 | 47 | 80 | |
| GenBank mRNA | CN837307.1 | - | 1 - 2 | 52 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001131054.1 transcripts.
Set Membership: |
|

Pttg1
Genomic Map