Assay Details
| Gene Symbol: Atp10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176999.3 | NP_795973.2 | 20 - 21 | 3742 | 102 | |
| GenBank mRNA | AK220352.1 | - | 10 - 11 | 1888 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp10b
Genomic Map