Assay Details
| Gene Symbol: Atl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178628.5 | NP_848743.1 | 1 - 2 | 243 | 78 | |
| GenBank mRNA | AK050339.1 | - | 1 - 2 | 80 | 78 | |
| AK147311.1 | - | 1 - 2 | 735 | 78 | ||
| CJ063033.1 | - | 1 - 2 | 156 | 78 | ||
| CX236218.1 | - | 1 - 2 | 243 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK050339.1, NM_178628.5 transcripts.
Set Membership: |
|

Atl1
Genomic Map