Assay Details
| Gene Symbol: Atl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178628.5 | NP_848743.1 | 3 - 4 | 673 | 101 | |
| GenBank mRNA | AK046919.1 | - | 2 - 3 | 403 | 101 | |
| AK050339.1 | - | 3 - 4 | 510 | 101 | ||
| AK140185.1 | - | 2 - 3 | 417 | 101 | ||
| AK147311.1 | - | 3 - 4 | 1165 | 101 | ||
| BC050784.1 | - | 2 - 3 | 402 | 6595181 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atl1
Genomic Map