Assay Details
| Gene Symbol: 5430435G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145509.2 | NP_663484.1 | 2 - 3 | 209 | 114 | |
| GenBank mRNA | AB232641.1 | - | 1 - 2 | 53 | 114 | |
| AK036056.1 | - | 2 - 3 | 198 | 114 | ||
| AK079999.1 | - | 2 - 3 | 177 | 114 | ||
| CJ137398.1 | - | 2 - 3 | 128 | 114 | ||
| JX431303.1 | - | 1 - 2 | 53 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

5430435G22Rik
Genomic Map