Assay Details
| Gene Symbol: Rassf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018750.3 | NP_061220.2 | 1 - 2 | 492 | 114 | |
| GenBank mRNA | AK005059.2 | - | 1 - 2 | 488 | 114 | |
| AK029321.1 | - | 1 - 2 | 494 | 115 | ||
| AK155534.1 | - | 1 - 2 | 488 | 114 | ||
| BC089605.1 | - | 1 - 2 | 494 | 30121804 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rassf5
Genomic Map