Assay Details
| Gene Symbol: Rassf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018750.3 | NP_061220.2 | 2 - 3 | 613 | 72 | |
| GenBank mRNA | AK029321.1 | - | 2 - 3 | 615 | 72 | |
| AK155534.1 | - | 2 - 3 | 609 | 72 | ||
| BC089605.1 | - | 2 - 3 | 615 | 30121804 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rassf5
Genomic Map