Assay Details
| Gene Symbol: Mecom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007963.2 | NP_031989.2 | 2 - 3 | 790 | 90 | |
| GenBank mRNA | AK033517.1 | - | 2 - 3 | 224 | 90 | |
| AK051098.1 | - | 2 - 3 | 647 | 90 | ||
| AK085548.1 | - | 2 - 3 | 791 | 90 | ||
| BC076620.1 | - | 2 - 3 | 742 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK051098.1, NM_007963.2 transcripts.
Set Membership: |
|

Mecom
Genomic Map