Assay Details
| Gene Symbol: Larp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016857.1 | - | 724 | 106 | ||
| AK136175.1 | - | 746 | 106 | |||
| BC100361.1 | - | 286 | 106 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Larp1b| Gene Symbol: Larp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016857.1 | - | 724 | 106 | ||
| AK136175.1 | - | 746 | 106 | |||
| BC100361.1 | - | 286 | 106 | |||
Set Membership: |
|