Assay Details
Gene Symbol: Fgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY027549.1 | - | 1 - 2 | 194 | 63 | |
AY027559.1 | - | 1 - 2 | 98 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY027549.1 | - | 1 - 2 | 194 | 63 | |
AY027559.1 | - | 1 - 2 | 98 | 63 |
Set Membership: |
![]() ![]() ![]() ![]() |