Assay Details
| Gene Symbol: 4932438A13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172679.2 | NP_766267.2 | 78 - 79 | 13810 | 116 | |
| GenBank mRNA | AK052702.1 | - | 10 - 11 | 1628 | 116 | |
| AK173100.1 | - | 22 - 23 | 3445 | 116 | ||
| BC027125.1 | - | 5 - 6 | 842 | 116 | ||
| BC079623.1 | - | 19 - 20 | 3180 | 116 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4932438A13Rik
Genomic Map