Assay Details
| Gene Symbol: Mtfr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253390.1 | NP_001240319.1 | 3 - 4 | 299 | 154 | |
| NM_001253391.1 | NP_001240320.1 | 3 - 4 | 461 | 154 | ||
| NM_026182.5 | NP_080458.1 | 3 - 4 | 309 | 154 | ||
| GenBank mRNA | AF354708.1 | - | 2 - 3 | 260 | 154 | |
| AK004846.1 | - | 3 - 4 | 287 | 154 | ||
| AK006952.1 | - | 3 - 4 | 461 | 154 | ||
| AK009195.2 | - | 3 - 4 | 275 | 154 | ||
| AK018048.1 | - | 3 - 4 | 291 | 154 | ||
| AK075822.1 | - | 3 - 4 | 285 | 154 | ||
| AK075992.1 | - | 3 - 4 | 309 | 154 | ||
| AK161251.1 | - | 3 - 4 | 282 | 154 | ||
| BC019971.1 | - | 3 - 4 | 257 | 4159093 | 154 | |
| DQ369744.1 | - | 3 - 4 | 420 | 154 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mtfr1
Genomic Map