Assay Details
| Gene Symbol: Gnb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013531.4 | NP_038559.2 | 1 - 2 | 100 | 73 | |
| GenBank mRNA | AK036816.1 | - | 1 - 2 | 102 | 73 | |
| AK044119.1 | - | 1 - 2 | 94 | 73 | ||
| AK152227.1 | - | 1 - 2 | 91 | 73 | ||
| AK165084.1 | - | 1 - 2 | 91 | 73 | ||
| AK167416.1 | - | 1 - 2 | 133 | 73 | ||
| AK168302.1 | - | 1 - 2 | 56 | 73 | ||
| AK169752.1 | - | 1 - 2 | 91 | 73 | ||
| BC028753.1 | - | 1 - 2 | 71 | 1348753 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK036816.1, AK152227.1, AK167416.1, AK168302.1, BC028753.1, NM_013531.4 transcripts.
Set Membership: |
|

Gnb4
Genomic Map