Assay Details
| Gene Symbol: Gnb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013531.4 | NP_038559.2 | 3 - 4 | 230 | 63 | |
| GenBank mRNA | AF277893.1 | - | 2 - 3 | 94 | 63 | |
| AF501886.1 | - | 2 - 3 | 94 | 63 | ||
| AK036816.1 | - | 3 - 4 | 232 | 63 | ||
| AK098139.1 | - | 3 - 4 | 163 | 63 | ||
| AK152227.1 | - | 3 - 4 | 221 | 63 | ||
| AK155309.1 | - | 3 - 4 | 289 | 63 | ||
| AK158965.1 | - | 4 - 5 | 460 | 63 | ||
| AK165084.1 | - | 3 - 4 | 221 | 63 | ||
| AK167416.1 | - | 3 - 4 | 263 | 63 | ||
| AK168302.1 | - | 3 - 4 | 186 | 63 | ||
| AK169752.1 | - | 3 - 4 | 221 | 63 | ||
| BC028753.1 | - | 3 - 4 | 201 | 1348753 | 63 | |
| M63658.1 | - | 2 - 3 | 134 | 63 | ||
| M87286.1 | - | 2 - 3 | 134 | 63 | ||
| S86124.1 | - | 2 - 3 | 134 | 63 | ||
More Information
Set Membership: |
|

Gnb4
Genomic Map