Assay Details
| Gene Symbol: Kcnmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028231.2 | NP_082507.1 | 2 - 3 | 321 | 80 | |
| GenBank mRNA | BC046227.1 | - | 2 - 3 | 321 | 5703879 | 80 |
| BC058957.1 | - | 2 - 3 | 321 | 5703879 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC046227.1, BC058957.1, NM_028231.2 transcripts.
Set Membership: |
|

Kcnmb2
Genomic Map