Assay Details
| Gene Symbol: Kcnmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012400.1 | - | 2 - 3 | 171 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK012400.1 transcripts.
Set Membership: |
|

Kcnmb2
Genomic Map