Assay Details
| Gene Symbol: Cpb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029706.1 | NP_083982.1 | 1 - 2 | 147 | 64 | |
| GenBank mRNA | BC138759.1 | - | 1 - 2 | 105 | 8861781 | 64 |
| BC138762.1 | - | 1 - 2 | 105 | 8861784 | 64 | |
| BQ831426.1 | - | 1 - 2 | 163 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpb1
Genomic Map