Assay Details
| Gene Symbol: Cpb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029706.1 | NP_083982.1 | 4 - 5 | 448 | 75 | |
| GenBank mRNA | AK003061.1 | - | 4 - 5 | 387 | 75 | |
| AK008678.1 | - | 1 - 2 | 120 | 75 | ||
| BC138759.1 | - | 4 - 5 | 406 | 8861781 | 75 | |
| BC138762.1 | - | 4 - 5 | 406 | 8861784 | 75 | |
| BQ831426.1 | - | 4 - 5 | 462 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpb1
Genomic Map