Assay Details
| Gene Symbol: Anxa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009673.2 | NP_033803.1 | 10 - 11 | 1056 | 151 | |
| GenBank mRNA | AK147740.1 | - | 10 - 11 | 835 | 151 | |
| AK152185.1 | - | 10 - 11 | 891 | 151 | ||
| AK153388.1 | - | 10 - 11 | 837 | 151 | ||
| AK153476.1 | - | 10 - 11 | 820 | 151 | ||
| D63423.1 | - | 10 - 11 | 794 | 151 | ||
| U29396.1 | - | 10 - 11 | 813 | 151 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anxa5
Genomic Map