Assay Details
| Gene Symbol: Acad9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172678.3 | NP_766266.3 | 10 - 11 | 1150 | 72 | |
| GenBank mRNA | AK028571.1 | - | 9 - 10 | 1864 | 72 | |
| AK031820.1 | - | 10 - 11 | 1082 | 72 | ||
| AK075984.1 | - | 10 - 11 | 1069 | 72 | ||
| AK145423.1 | - | 10 - 11 | 1058 | 72 | ||
| AK206153.1 | - | 4 - 5 | 364 | 72 | ||
| AK217282.1 | - | 4 - 5 | 374 | 72 | ||
More Information
Set Membership: |
|

Acad9
Genomic Map