Assay Details
| Gene Symbol: Elf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291059.1 | NP_001277988.1 | 6 - 7 | 974 | 64 | |
| NM_001291062.1 | NP_001277991.1 | 3 - 4 | 569 | 64 | ||
| NM_001291063.1 | NP_001277992.1 | 3 - 4 | 605 | 64 | ||
| NM_023502.2 | NP_075991.1 | 6 - 7 | 1010 | 64 | ||
| GenBank mRNA | AF256216.1 | - | 5 - 6 | 695 | 64 | |
| AF256217.1 | - | 5 - 6 | 731 | 64 | ||
| AF256218.1 | - | 3 - 4 | 449 | 64 | ||
| AF256219.1 | - | 3 - 4 | 485 | 64 | ||
| AK088422.1 | - | 3 - 4 | 570 | 64 | ||
| AK134180.1 | - | 1 - 2 | 40 | 64 | ||
| AK146819.1 | - | 6 - 7 | 975 | 64 | ||
| AK160118.1 | - | 6 - 7 | 880 | 64 | ||
| BC051937.1 | - | 5 - 6 | 781 | 64 | ||
| BC092057.1 | - | 5 - 6 | 753 | 4016339 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Elf2
Genomic Map