Assay Details
| Gene Symbol: Elf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291059.1 | NP_001277988.1 | 5 - 6 | 826 | 92 | |
| NM_001291062.1 | NP_001277991.1 | 2 - 3 | 421 | 92 | ||
| GenBank mRNA | AF256216.1 | - | 4 - 5 | 547 | 92 | |
| AF256218.1 | - | 2 - 3 | 301 | 92 | ||
| AK088422.1 | - | 2 - 3 | 422 | 92 | ||
| AK139199.1 | - | 3 - 4 | 426 | 92 | ||
| AK146819.1 | - | 5 - 6 | 827 | 92 | ||
| AK160118.1 | - | 5 - 6 | 732 | 92 | ||
| BC027739.1 | - | 6 - 7 | 945 | 92 | ||
| BC069901.1 | - | 5 - 6 | 822 | 92 | ||
| BC092057.1 | - | 4 - 5 | 605 | 4016339 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Elf2
Genomic Map