Assay Details
| Gene Symbol: Skil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011386.3 | NP_035516.2 | 4 - 5 | 2145 | 112 | |
| GenBank mRNA | AB221599.1 | - | 3 - 4 | 1422 | 112 | |
| CB519078.1 | - | 1 - 2 | 136 | 112 | ||
| U10532.1 | - | 3 - 4 | 1916 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Skil
Genomic Map