Assay Details
| Gene Symbol: Sec62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027016.2 | NP_081292.1 | 6 - 7 | 1082 | 83 | |
| GenBank mRNA | AK088093.1 | - | 6 - 7 | 631 | 83 | |
| AK090078.1 | - | 6 - 7 | 632 | 83 | ||
| BC023720.1 | - | 3 - 4 | 297 | 83 | ||
| BC053504.1 | - | 6 - 7 | 620 | 83 | ||
| BC067202.1 | - | 6 - 7 | 617 | 6332701 | 83 | |
| BC071243.1 | - | 6 - 7 | 633 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec62
Genomic Map