Assay Details
| Gene Symbol: Hspa4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011020.3 | NP_035150.3 | 6 - 7 | 726 | 83 | |
| GenBank mRNA | AK144225.1 | - | 6 - 7 | 763 | 83 | |
| AK163459.1 | - | 7 - 8 | 898 | 83 | ||
| BC057002.1 | - | 6 - 7 | 726 | 5702602 | 83 | |
| BC110662.1 | - | 6 - 7 | 746 | 4511154 | 83 | |
| U23921.1 | - | 6 - 7 | 843 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hspa4l
Genomic Map