Assay Details
| Gene Symbol: Hspa4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011020.3 | NP_035150.3 | 8 - 9 | 1047 | 85 | |
| GenBank mRNA | AB001926.1 | - | 8 - 9 | 1013 | 85 | |
| AK144225.1 | - | 8 - 9 | 1084 | 85 | ||
| AK163459.1 | - | 9 - 10 | 1219 | 85 | ||
| BC012712.1 | - | 8 - 9 | 1116 | 4190161 | 85 | |
| BC057002.1 | - | 8 - 9 | 1047 | 5702602 | 85 | |
| BC110662.1 | - | 8 - 9 | 1067 | 4511154 | 85 | |
| D49482.1 | - | 8 - 9 | 1041 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hspa4l
Genomic Map