Assay Details
| Gene Symbol: Pcdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098172.1 | NP_001091642.1 | 4 - 5 | 3954 | 67 | |
| GenBank mRNA | AK081621.1 | - | 4 - 5 | 3953 | 67 | |
| AK081977.1 | - | 4 - 5 | 3950 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcdh10
Genomic Map