Assay Details
| Gene Symbol: Wwtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133784.3 | NP_598545.2 | 1 - 2 | 274 | 79 | |
| GenBank mRNA | AJ299430.1 | - | 1 - 2 | 209 | 79 | |
| AK009983.1 | - | 1 - 2 | 207 | 79 | ||
| AK011504.1 | - | 1 - 2 | 79 | |||
| AK142720.1 | - | 1 - 2 | 199 | 79 | ||
| AK213529.1 | - | 1 - 2 | 85 | 79 | ||
| BC004640.1 | - | 1 - 2 | 195 | 3157246 | 79 | |
| BC014727.1 | - | 1 - 2 | 226 | 4487316 | 79 | |
| BY145519.1 | - | 1 - 2 | 275 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ299430.1, AK142720.1, BC004640.1, BC014727.1, NM_133784.3 transcripts.
Set Membership: |
|

Wwtr1
Genomic Map