Assay Details
| Gene Symbol: Wwtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168281.1 | NP_001161753.1 | 5 - 6 | 1093 | 60 | |
| NM_133784.3 | NP_598545.2 | 5 - 6 | 1172 | 60 | ||
| GenBank mRNA | AJ299430.1 | - | 5 - 6 | 1107 | 60 | |
| AK009983.1 | - | 5 - 6 | 1103 | 60 | ||
| AK011504.1 | - | 5 - 6 | 1146 | 60 | ||
| AK142299.1 | - | 5 - 6 | 1094 | 60 | ||
| AK142720.1 | - | 5 - 6 | 1097 | 60 | ||
| BC004640.1 | - | 5 - 6 | 1093 | 3157246 | 60 | |
| BC014727.1 | - | 5 - 6 | 1124 | 4487316 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wwtr1
Genomic Map