Assay Details
| Gene Symbol: P2ry12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027571.3 | NP_081847.3 | 2 - 3 | 196 | 74 | |
| GenBank mRNA | AK013804.1 | - | 2 - 3 | 197 | 74 | |
| BC027381.1 | - | 1 - 2 | 56 | 4947057 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK013804.1, BC027381.1, NM_027571.3 transcripts.
Set Membership: |
|

P2ry12
Genomic Map