Assay Details
| Gene Symbol: Supt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019995.3 | NP_064379.1 | 11 - 12 | 1054 | 87 | |
| GenBank mRNA | AF139179.1 | - | 11 - 12 | 1058 | 87 | |
| AK050619.1 | - | 12 - 13 | 1078 | 87 | ||
| AK083379.1 | - | 12 - 13 | 1078 | 87 | ||
| AK207764.1 | - | 1 - 2 | 58 | 87 | ||
| BC052702.1 | - | 11 - 12 | 1043 | 6400922 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Supt20
Genomic Map