Assay Details
| Gene Symbol: Supt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019995.3 | NP_064379.1 | 17 - 18 | 1515 | 80 | |
| GenBank mRNA | AF139179.1 | - | 17 - 18 | 1519 | 80 | |
| BC052702.1 | - | 17 - 18 | 1504 | 6400922 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Supt20
Genomic Map